Product Details

SNP ID
rs147390697
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:152769948 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGATGCAATGTATCAACTGCATCTA[A/G]TAGTGAAAATGTAAATTCTTTGCTA
Phenotype
MIM: 605928
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ARFIP1 PubMed Links

Gene Details

Gene
ARFIP1
Gene Name
ADP ribosylation factor interacting protein 1
There are no transcripts associated with this gene.

Gene
TIGD4
Gene Name
tigger transposable element derived 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_145720.3 1878 Silent Mutation NP_663772.1
XM_005262807.4 1878 Silent Mutation XP_005262864.1

View Full Product Details