Product Details

SNP ID
rs150910491
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:5016868 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGAACGAGTTCATGATGGTGTACA[A/G]CTTCCGTGCTTTGTCCTTCAAGGAA
Phenotype
MIM: 607930
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CYTL1 PubMed Links

Gene Details

Gene
CYTL1
Gene Name
cytokine like 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018659.2 320 Silent Mutation CTG,TTG L99L NP_061129.1
XM_017008299.1 320 Missense Mutation GCT,GTT A137V XP_016863788.1

View Full Product Details