Product Details
- SNP ID
-
rs151077178
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.4:163471883 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TTCATTAAAGTAAGTGTTACGGCTG[C/T]TATAATGTGTCTGGTACTGATTCCA
- Phenotype
-
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
TKTL2
PubMed Links
Gene Details
- Gene
- TKTL2
- Gene Name
- transketolase like 2
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_032136.4 |
2013 |
Missense Mutation |
|
|
NP_115512.3 |
View Full Product Details