Product Details
- SNP ID
-
rs151152950
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.4:48490644 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GTGAGCCCGCGCATCGACGCCGCGG[C/T]ACAGTGCTCGCTGGGGAGGCGCACG
- Phenotype
-
MIM: 607520
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
FRYL
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs74929644] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- FRYL
- Gene Name
- FRY like transcription coactivator
There are no transcripts associated with this gene.
- Gene
- SLC10A4
- Gene Name
- solute carrier family 10 member 4
There are no transcripts associated with this gene.
- Gene
- ZAR1
- Gene Name
- zygote arrest 1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_175619.2 |
393 |
Missense Mutation |
GCA,GTA |
A118V |
NP_783318.1 |
View Full Product Details