Product Details

SNP ID
rs138087009
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:156920494 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTGTTTCTGCGAACAATAGGTTTCC[A/G]TGAGTTTCCCTGAAAAGACAAGGCC
Phenotype
MIM: 610096
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TIMD4 PubMed Links

Gene Details

Gene
TIMD4
Gene Name
T-cell immunoglobulin and mucin domain containing 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001146726.1 995 Missense Mutation ACG,ATG T313M NP_001140198.1
NM_138379.2 995 Missense Mutation ACG,ATG T341M NP_612388.2
XM_011534694.2 995 Intron XP_011532996.1
XM_017010021.1 995 Missense Mutation ACG,ATG T286M XP_016865510.1

View Full Product Details