Product Details

SNP ID
rs138616616
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:76953648 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATCCTTTCCTGCTCTTCAGCGCCAA[C/T]CTGAAGCGGGAGCTGGCTGGGGAGC
Phenotype
MIM: 122559 MIM: 610103
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CRHBP PubMed Links

Gene Details

Gene
CRHBP
Gene Name
corticotropin releasing hormone binding protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001882.3 409 Silent Mutation AAC,AAT N43N NP_001873.2
Gene
S100Z
Gene Name
S100 calcium binding protein Z
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_130772.3 409 Intron NP_570128.2
XM_011543239.2 409 Intron XP_011541541.1
XM_011543240.2 409 Intron XP_011541542.1
XM_011543241.2 409 Intron XP_011541543.1
XM_011543244.2 409 Intron XP_011541546.1
XM_011543245.2 409 Intron XP_011541547.1
XM_017009171.1 409 Intron XP_016864660.1
XM_017009172.1 409 Intron XP_016864661.1
XM_017009173.1 409 Intron XP_016864662.1

View Full Product Details