Product Details

SNP ID
rs140359856
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:141319301 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCTGTCTCTCCAGATCTTCCTCCGT[C/G]TCAATGATGTTTATATCTTCTTCAT
Phenotype
MIM: 600573
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
TAF7 PubMed Links

Gene Details

Gene
TAF7
Gene Name
TATA-box binding protein associated factor 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005642.2 1484 Silent Mutation GAC,GAG D248E NP_005633.2

View Full Product Details