Product Details

SNP ID
rs141381751
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:79111970 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCTCCTTCCCCTCCCACCTGCTCT[G/T]TATCCCAGCCTCTGGGAGCGCAGAG
Phenotype
MIM: 602888
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
BHMT PubMed Links

Gene Details

Gene
BHMT
Gene Name
betaine--homocysteine S-methyltransferase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001713.2 Intron NP_001704.2

View Full Product Details