Product Details

SNP ID
rs144765111
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:141319945 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGTCCTTGAGGTTGACATGACCAG[A/C]CTGTACTGCCCTTCTCACAGTAGAG
Phenotype
MIM: 600573
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
TAF7 PubMed Links

Gene Details

Gene
TAF7
Gene Name
TATA-box binding protein associated factor 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005642.2 840 Missense Mutation GCT,TCT A34S NP_005633.2

View Full Product Details