Product Details

SNP ID
rs145318066
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:129748258 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCCTGCAGCTTGACGTAAAGTCTT[C/T]AACGAGGAGCTCAGCCCTCCTTCAG
Phenotype
MIM: 607513
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ADAMTS19 PubMed Links

Gene Details

Gene
ADAMTS19
Gene Name
ADAM metallopeptidase with thrombospondin type 1 motif 19
There are no transcripts associated with this gene.

Gene
KIAA1024L
Gene Name
KIAA1024 like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001257308.1 68 Missense Mutation TCA,TTA S23L NP_001244237.1

View Full Product Details