Product Details

SNP ID
rs146158267
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:91369109 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAAAACTTTATTTGATAAAGTTTTA[G/T]ACATTTAAAGTTTTATCACATTTTG
Phenotype
MIM: 612464
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
ARRDC3 PubMed Links

Gene Details

Gene
ARRDC3
Gene Name
arrestin domain containing 3
There are no transcripts associated with this gene.

View Full Product Details