Product Details

SNP ID
rs148011374
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:180911411 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GACATCCAGCTACTCTGTCAGTCCT[C/T]GGGCTGGTTCCCCCGGCCCACAGCG
Phenotype
MIM: 615606
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
BTNL8 PubMed Links

Gene Details

Gene
BTNL8
Gene Name
butyrophilin like 8
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001040462.2 509 Missense Mutation TCG,TTG S157L NP_001035552.1
NM_001159707.1 509 Missense Mutation TCG,TTG S41L NP_001153179.1
NM_001159708.1 509 Missense Mutation TCG,TTG S157L NP_001153180.1
NM_001159709.1 509 Missense Mutation TCG,TTG S32L NP_001153181.1
NM_001159710.1 509 UTR 5 NP_001153182.1
NM_024850.2 509 Missense Mutation TCG,TTG S157L NP_079126.1
XM_011534649.1 509 Missense Mutation TCG,TTG S157L XP_011532951.1
XM_011534650.2 509 Missense Mutation TCG,TTG S107L XP_011532952.1
XM_011534651.2 509 Missense Mutation TCG,TTG S46L XP_011532953.1
XM_011534652.2 509 Missense Mutation TCG,TTG S38L XP_011532954.1
XM_011534653.2 509 Intron XP_011532955.1

View Full Product Details