Product Details

SNP ID
rs148225175
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:69509711 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAACTGCTCAGTCTTCTGGATCTCT[A/G]TATGGTTCACAAATATATGCCCTCT
Phenotype
MIM: 602876
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
OCLN PubMed Links

Gene Details

Gene
OCLN
Gene Name
occludin
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001205254.1 1057 Silent Mutation CTA,CTG L207L NP_001192183.1
NM_001205255.1 1057 Intron NP_001192184.1
NM_002538.3 1057 Silent Mutation CTA,CTG L207L NP_002529.1
XM_017008913.1 1057 Silent Mutation CTA,CTG L207L XP_016864402.1
XM_017008914.1 1057 Silent Mutation CTA,CTG L207L XP_016864403.1

View Full Product Details