Product Details
- SNP ID
-
rs148336496
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.5:97094894 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CGTAGTTCTTGTCCTGCTTTCCTGG[C/T]TTCCTCCAACTGCCTCAGGGCCATT
- Phenotype
-
MIM: 610466
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
LIX1
PubMed Links
Gene Details
- Gene
- LIX1
- Gene Name
- limb and CNS expressed 1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_153234.4 |
943 |
Missense Mutation |
ACC,GCC |
T235A |
NP_694966.3 |
View Full Product Details