Product Details

SNP ID
rs148587063
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:141320018 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGATAAACTGGCTCTCCAGTTCGTG[A/G]GGAGCATCATCTTTGCTTTTACTCA
Phenotype
MIM: 600573
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TAF7 PubMed Links

Gene Details

Gene
TAF7
Gene Name
TATA-box binding protein associated factor 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005642.2 767 Silent Mutation CCC,CCT P9P NP_005633.2

View Full Product Details