Product Details

SNP ID
rs149079138
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:141319185 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGTCTCTTGGAGCTTGCCTTTCATG[C/T]TGTCAATCTGCTTCTGAATTCCCAT
Phenotype
MIM: 600573
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TAF7 PubMed Links

Gene Details

Gene
TAF7
Gene Name
TATA-box binding protein associated factor 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005642.2 1600 Missense Mutation AAC,AGC N287S NP_005633.2

View Full Product Details