Product Details
- SNP ID
-
rs150936557
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.5:132074859 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CTGGAGCTGTACAAGCAGGGCCTGC[A/G]GGGCAGCCTCACCAAGCTCAAGGGC
- Phenotype
-
MIM: 138960
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
CSF2
PubMed Links
Gene Details
- Gene
- CSF2
- Gene Name
- colony stimulating factor 2
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_000758.3 |
283 |
Missense Mutation |
CAG,CGG |
Q84R |
NP_000749.2 |
View Full Product Details