Product Details

SNP ID
rs17194460
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:30163769 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGGCCGCTGGAGGATGCGGTGACC[A/G]TTCCCTGTGGACACACCTTCTGCCG
Phenotype
MIM: 605701
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TRIM10 PubMed Links

Gene Details

Gene
TRIM10
Gene Name
tripartite motif containing 10
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006778.3 564 Intron NP_006769.2
NM_052828.2 564 Intron NP_439893.2
XM_011514221.1 564 Intron XP_011512523.1
XM_011514222.2 564 Intron XP_011512524.1
XM_011514223.2 564 Intron XP_011512525.1
XM_011514225.1 564 Intron XP_011512527.1
Gene
TRIM15
Gene Name
tripartite motif containing 15
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_033229.2 564 Missense Mutation ATT,GTT I29V NP_150232.2
XM_011514987.1 564 Intron XP_011513289.1
XM_011514988.2 564 UTR 5 XP_011513290.1

View Full Product Details