Product Details

SNP ID
rs138921957
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:57178300 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AATGGAAAAATTGTAGACAAAAGAC[A/G]TGTATATGAGTTTAAGGATAGCAAT
Phenotype
MIM: 603882 MIM: 606144 MIM: 615708
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
BAG2 PubMed Links

Gene Details

Gene
BAG2
Gene Name
BCL2 associated athanogene 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004282.3 Intron NP_004273.1
XM_005249490.3 Intron XP_005249547.1
XM_011514998.2 Intron XP_011513300.1
XM_011514999.2 Intron XP_011513301.1
Gene
LOC101927211
Gene Name
uncharacterized LOC101927211
There are no transcripts associated with this gene.

Gene
RAB23
Gene Name
RAB23, member RAS oncogene family
There are no transcripts associated with this gene.

Gene
ZNF451
Gene Name
zinc finger protein 451
There are no transcripts associated with this gene.

View Full Product Details