Product Details

SNP ID
rs139799684
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:12723814 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTAAATGCTAGAGTCCACTATGGAC[C/T]AATTAAATCAGAATCTCTGGGGATA
Phenotype
MIM: 608723
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PHACTR1 PubMed Links

Gene Details

Gene
PHACTR1
Gene Name
phosphatase and actin regulator 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001242648.2 Intron NP_001229577.1
NM_001322308.1 Intron NP_001309237.1
NM_001322309.1 Intron NP_001309238.1
NM_001322310.1 Intron NP_001309239.1
NM_001322311.1 Intron NP_001309240.1
NM_001322312.1 Intron NP_001309241.1
NM_001322313.1 Intron NP_001309242.1
NM_001322314.1 Intron NP_001309243.1
NM_030948.3 Intron NP_112210.1
XM_005248934.2 Intron XP_005248991.1
XM_017010452.1 Intron XP_016865941.1
XM_017010454.1 Intron XP_016865943.1
XM_017010455.1 Intron XP_016865944.1
XM_017010456.1 Intron XP_016865945.1
XM_017010457.1 Intron XP_016865946.1
XM_017010458.1 Intron XP_016865947.1
XM_017010459.1 Intron XP_016865948.1
XM_017010460.1 Intron XP_016865949.1
XM_017010461.1 Intron XP_016865950.1
XM_017010462.1 Intron XP_016865951.1
XM_017010463.1 Intron XP_016865952.1
XM_017010464.1 Intron XP_016865953.1
XM_017010465.1 Intron XP_016865954.1
XM_017010466.1 Intron XP_016865955.1
XM_017010467.1 Intron XP_016865956.1
XM_017010469.1 Intron XP_016865958.1

View Full Product Details