Product Details

SNP ID
rs140347150
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:26240614 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCGCGGTGTTCTTAAGGTGTTCCT[A/G]GAGAATGTGATACGGGACGCCGTAA
Phenotype
MIM: 142210 MIM: 602816 MIM: 602824 MIM: 602832
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
HIST1H1D PubMed Links

Gene Details

Gene
HIST1H1D
Gene Name
histone cluster 1, H1d
There are no transcripts associated with this gene.

Gene
HIST1H3F
Gene Name
histone cluster 1, H3f
There are no transcripts associated with this gene.

Gene
HIST1H4F
Gene Name
histone cluster 1, H4f
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003540.3 189 Silent Mutation CTA,CTG L63L NP_003531.1
Gene
HIST1H4G
Gene Name
histone cluster 1, H4g
There are no transcripts associated with this gene.

View Full Product Details