Product Details

SNP ID
rs145208127
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:24503412 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCTGGTCCTCAAGCAGCCCATAGG[A/C]GTGGCTGCAGTCATCACCCCGGTAG
Phenotype
MIM: 610045 MIM: 602515
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
ALDH5A1 PubMed Links

Gene Details

Gene
ALDH5A1
Gene Name
aldehyde dehydrogenase 5 family member A1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001080.3 616 Silent Mutation GGA,GGC G196G NP_001071.1
NM_170740.1 616 Silent Mutation GGA,GGC G196G NP_733936.1
Gene
GPLD1
Gene Name
glycosylphosphatidylinositol specific phospholipase D1
There are no transcripts associated with this gene.

View Full Product Details