Product Details

SNP ID
rs145599850
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:109979571 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGGCTTCCTCGTGGCCTCCTTCGC[A/C]GCCTCTGTCAGCAGCCTGCTGGCCA
Phenotype
MIM: 600553
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
GPR6 PubMed Links

Gene Details

Gene
GPR6
Gene Name
G protein-coupled receptor 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001286099.1 798 Silent Mutation GCA,GCC A168A NP_001273028.1
NM_005284.4 798 Silent Mutation GCA,GCC A153A NP_005275.1

View Full Product Details