Product Details

SNP ID
rs145678004
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:116880552 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTGTTCCTTTTTCTTAGGCTTGAAG[A/G]GAATTACATTGTATGTGAAGGAGTT
Phenotype
MIM: 612659
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
RFX6 PubMed Links

Gene Details

Gene
RFX6
Gene Name
regulatory factor X6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_173560.3 452 Missense Mutation GAG,GGG E130G NP_775831.2
XM_011535589.1 452 Missense Mutation GAG,GGG E130G XP_011533891.1
XM_017010477.1 452 Missense Mutation GAG,GGG E4G XP_016865966.1

View Full Product Details