Product Details
- SNP ID
-
rs147439442
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.6:24503366 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CGTGTTTACGGAGACATTATCCACA[C/T]CCCGGCAAAGGACAGGCGGGCCCTG
- Phenotype
-
MIM: 610045
MIM: 602515
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
ALDH5A1
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs2760118,rs3765310] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- ALDH5A1
- Gene Name
- aldehyde dehydrogenase 5 family member A1
- Gene
- GPLD1
- Gene Name
- glycosylphosphatidylinositol specific phospholipase D1
There are no transcripts associated with this gene.
View Full Product Details