Product Details

SNP ID
rs147898099
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:54308687 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AATCACACCGTTTTGCAAGGTACTC[C/G]ATTCAAAAGAGCCATTTTCCAAGGG
Phenotype
MIM: 606749
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
TINAG PubMed Links

Gene Details

Gene
TINAG
Gene Name
tubulointerstitial nephritis antigen
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014464.3 283 Missense Mutation CCA,CGA P46R NP_055279.3
XM_006715062.2 283 Missense Mutation CCA,CGA P28R XP_006715125.1
XM_011514497.2 283 Missense Mutation CCA,CGA P46R XP_011512799.1
XM_017010745.1 283 Missense Mutation CCA,CGA P46R XP_016866234.1
XM_017010746.1 283 Intron XP_016866235.1
XM_017010747.1 283 Intron XP_016866236.1
XM_017010748.1 283 Intron XP_016866237.1

View Full Product Details