Product Details

SNP ID
rs148582871
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:169702814 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCGGCGCCCAGTGGGCATCGGCGTC[A/T]ATGGACACCCCTCCCACCTGGAGAG
Phenotype
MIM: 616987 MIM: 613069
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
C6orf120 PubMed Links

Gene Details

Gene
C6orf120
Gene Name
chromosome 6 open reading frame 120
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001029863.2 703 Missense Mutation AAT,TAT N119Y NP_001025034.1
NM_001317342.1 703 Missense Mutation AAT,TAT N138Y NP_001304271.1
Gene
PHF10
Gene Name
PHD finger protein 10
There are no transcripts associated with this gene.

Gene
WDR27
Gene Name
WD repeat domain 27
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001202550.1 703 Intron NP_001189479.1
NM_182552.4 703 Intron NP_872358.4
XM_011535682.2 703 Intron XP_011533984.1
XM_011535684.2 703 Intron XP_011533986.1
XM_011535685.2 703 Intron XP_011533987.1
XM_011535687.2 703 Intron XP_011533989.1
XM_011535688.2 703 Intron XP_011533990.1
XM_011535689.2 703 Intron XP_011533991.1
XM_011535690.2 703 Intron XP_011533992.1
XM_011535691.2 703 Intron XP_011533993.1
XM_011535692.2 703 Intron XP_011533994.1
XM_011535693.2 703 Intron XP_011533995.1
XM_011535694.2 703 Intron XP_011533996.1
XM_011535696.2 703 Intron XP_011533998.1
XM_011535697.2 703 Intron XP_011533999.1
XM_017010658.1 703 Intron XP_016866147.1
XM_017010659.1 703 Intron XP_016866148.1
XM_017010660.1 703 Intron XP_016866149.1
XM_017010661.1 703 Intron XP_016866150.1
XM_017010662.1 703 Intron XP_016866151.1
XM_017010663.1 703 Intron XP_016866152.1
XM_017010664.1 703 Intron XP_016866153.1
XM_017010665.1 703 Intron XP_016866154.1
XM_017010666.1 703 Intron XP_016866155.1
XM_017010667.1 703 Intron XP_016866156.1
XM_017010668.1 703 Intron XP_016866157.1
XM_017010669.1 703 Intron XP_016866158.1
XM_017010670.1 703 Intron XP_016866159.1
XM_017010671.1 703 Intron XP_016866160.1
XM_017010672.1 703 Intron XP_016866161.1
XM_017010673.1 703 Intron XP_016866162.1
XM_017010674.1 703 Intron XP_016866163.1
XM_017010675.1 703 Intron XP_016866164.1

View Full Product Details