Product Details

SNP ID
rs149161868
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:43025289 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTCTGGAAATGTCAGCCAAGATCC[A/G]AGTACCATTTTTACGTCAGGTTGTT
Phenotype
MIM: 611248 MIM: 143170 MIM: 613475
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
KLHDC3 PubMed Links

Gene Details

Gene
KLHDC3
Gene Name
kelch domain containing 3
There are no transcripts associated with this gene.

Gene
MEA1
Gene Name
male-enhanced antigen 1
There are no transcripts associated with this gene.

Gene
RRP36
Gene Name
ribosomal RNA processing 36
There are no transcripts associated with this gene.

View Full Product Details