Product Details
- SNP ID
-
rs149689024
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.6:109099331 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AGCCAGTGAGCAGAGAAAAACCCAC[G/T]GTTCAGTAGGTGTTCTAGAAGCAAA
- Phenotype
-
MIM: 606103
- Polymorphism
- G/T, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
CEP57L1
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs201271917] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- CEP57L1
- Gene Name
- centrosomal protein 57 like 1
- Gene
- SESN1
- Gene Name
- sestrin 1
There are no transcripts associated with this gene.
View Full Product Details