Product Details

SNP ID
rs151116596
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:169702874 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GATGAAGGTGTACTACGACGGCACG[A/G]TCGAGCAGCACCCGTTCGGCGAGGC
Phenotype
MIM: 616987 MIM: 613069
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
C6orf120 PubMed Links

Gene Details

Gene
C6orf120
Gene Name
chromosome 6 open reading frame 120
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001029863.2 763 Missense Mutation ATC,GTC I139V NP_001025034.1
NM_001317342.1 763 Missense Mutation ATC,GTC I158V NP_001304271.1
Gene
PHF10
Gene Name
PHD finger protein 10
There are no transcripts associated with this gene.

Gene
WDR27
Gene Name
WD repeat domain 27
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001202550.1 763 Intron NP_001189479.1
NM_182552.4 763 Intron NP_872358.4
XM_011535682.2 763 Intron XP_011533984.1
XM_011535684.2 763 Intron XP_011533986.1
XM_011535685.2 763 Intron XP_011533987.1
XM_011535687.2 763 Intron XP_011533989.1
XM_011535688.2 763 Intron XP_011533990.1
XM_011535689.2 763 Intron XP_011533991.1
XM_011535690.2 763 Intron XP_011533992.1
XM_011535691.2 763 Intron XP_011533993.1
XM_011535692.2 763 Intron XP_011533994.1
XM_011535693.2 763 Intron XP_011533995.1
XM_011535694.2 763 Intron XP_011533996.1
XM_011535696.2 763 Intron XP_011533998.1
XM_011535697.2 763 Intron XP_011533999.1
XM_017010658.1 763 Intron XP_016866147.1
XM_017010659.1 763 Intron XP_016866148.1
XM_017010660.1 763 Intron XP_016866149.1
XM_017010661.1 763 Intron XP_016866150.1
XM_017010662.1 763 Intron XP_016866151.1
XM_017010663.1 763 Intron XP_016866152.1
XM_017010664.1 763 Intron XP_016866153.1
XM_017010665.1 763 Intron XP_016866154.1
XM_017010666.1 763 Intron XP_016866155.1
XM_017010667.1 763 Intron XP_016866156.1
XM_017010668.1 763 Intron XP_016866157.1
XM_017010669.1 763 Intron XP_016866158.1
XM_017010670.1 763 Intron XP_016866159.1
XM_017010671.1 763 Intron XP_016866160.1
XM_017010672.1 763 Intron XP_016866161.1
XM_017010673.1 763 Intron XP_016866162.1
XM_017010674.1 763 Intron XP_016866163.1
XM_017010675.1 763 Intron XP_016866164.1

View Full Product Details