Product Details

SNP ID
rs138629558
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:30654148 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCTCTGTCTTGGTCATTTCTAAAG[C/T]TGGCTCTACCCGGTGCCATTTAAGC
Phenotype
MIM: 602034
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CRHR2 PubMed Links

Gene Details

Gene
CRHR2
Gene Name
corticotropin releasing hormone receptor 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001202475.1 1881 Intron NP_001189404.1
NM_001202481.1 1881 Intron NP_001189410.1
NM_001202482.1 1881 Intron NP_001189411.1
NM_001202483.1 1881 Intron NP_001189412.1
NM_001883.4 1881 Intron NP_001874.2
XM_011515127.2 1881 Intron XP_011513429.1
XM_011515128.2 1881 UTR 3 XP_011513430.1
XM_011515129.2 1881 UTR 3 XP_011513431.1
XM_017011751.1 1881 Intron XP_016867240.1
XM_017011752.1 1881 Intron XP_016867241.1

View Full Product Details