Product Details

SNP ID
rs141364612
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:73736565 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATCTTCTTGCCAGCAACTCTTAGAC[C/T]AGGAAGCCTCGGATGGCAGCTATGA
Phenotype
MIM: 612545
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ABHD11 PubMed Links

Gene Details

Gene
ABHD11
Gene Name
abhydrolase domain containing 11
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001145364.2 1109 Silent Mutation CTA,CTG L257L NP_001138836.1
NM_001301058.1 1109 UTR 3 NP_001287987.1
NM_001321382.1 1109 Silent Mutation CTA,CTG L201L NP_001308311.1
NM_001321383.1 1109 UTR 3 NP_001308312.1
NM_148912.3 1109 Silent Mutation CTA,CTG L314L NP_683710.1
NM_148913.3 1109 Silent Mutation CTA,CTG L307L NP_683711.1
XM_006716144.2 1109 Silent Mutation CTA,CTG L201L XP_006716207.1
XM_011516590.2 1109 Silent Mutation CTA,CTG L293L XP_011514892.1
XM_011516591.1 1109 Silent Mutation CTA,CTG L201L XP_011514893.1
Gene
ABHD11-AS1
Gene Name
ABHD11 antisense RNA 1 (tail to tail)
There are no transcripts associated with this gene.

View Full Product Details