Product Details

SNP ID
rs142406481
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:44799264 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTTACAGCTGTTTGCAGACAAGGT[C/T]CCAAAGACAGCAGGTTGGTCCATTT
Phenotype
MIM: 123840
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PPIA PubMed Links

Gene Details

Gene
PPIA
Gene Name
peptidylprolyl isomerase A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001300981.1 176 Intron NP_001287910.1
NM_021130.4 176 Silent Mutation GTC,GTT V29V NP_066953.1

View Full Product Details