Product Details

SNP ID
rs142570247
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:98848615 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATGTCCTTTTGCTGAGTGGCATTCC[A/G]CAGGGTCATGTACATGATGAAGGCC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TMEM130 PubMed Links

Gene Details

Gene
TMEM130
Gene Name
transmembrane protein 130
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001134450.1 970 Silent Mutation NP_001127922.1
NM_001134451.1 970 Silent Mutation NP_001127923.1
NM_152913.2 970 Silent Mutation NP_690877.1

View Full Product Details