Product Details

SNP ID
rs143856570
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:152648877 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATGAGAAGGTTCTTCTGATGAGCTC[A/G]AGGCTTTCTGCATTATAGTTTGTGT
Phenotype
MIM: 600375
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
XRCC2 PubMed Links

Gene Details

Gene
XRCC2
Gene Name
X-ray repair cross complementing 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005431.1 694 Missense Mutation TCG,TTG S203L NP_005422.1

View Full Product Details