Product Details

SNP ID
rs145100726
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:102457893 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGCCCCGGGTAGAGGCCCATCCTC[C/T]GCACCACCTCCCGGCTGAAGCTGGG
Phenotype
MIM: 613302 MIM: 615167 MIM: 610929
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ALKBH4 PubMed Links

Gene Details

Gene
ALKBH4
Gene Name
alkB homolog 4, lysine demethylase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_017621.3 448 Missense Mutation CAG,CGG Q137R NP_060091.1
XM_005250464.2 448 Missense Mutation CAG,CGG Q64R XP_005250521.1
XM_005250465.2 448 UTR 5 XP_005250522.1
Gene
LRWD1
Gene Name
leucine rich repeats and WD repeat domain containing 1
There are no transcripts associated with this gene.

Gene
MIR5090
Gene Name
microRNA 5090
There are no transcripts associated with this gene.

Gene
ORAI2
Gene Name
ORAI calcium release-activated calcium modulator 2
There are no transcripts associated with this gene.

View Full Product Details