Product Details

SNP ID
rs147151212
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.7:140673530 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAGGGGGCCCCTGACGTTCTGAGTC[A/G]GCGAAGGGTCCGCTGCAGCGGGGCG
Phenotype
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
ADCK2 PubMed Links
Additional Information
For this assay, SNP(s) [rs2968558] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.

Gene Details

Gene
ADCK2
Gene Name
aarF domain containing kinase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_052853.3 603 Missense Mutation CAG,CGG Q67R NP_443085.2
XM_006716170.3 603 Missense Mutation CAG,CGG Q67R XP_006716233.1
XM_011516675.2 603 Missense Mutation CAG,CGG Q67R XP_011514977.1

View Full Product Details