Product Details

SNP ID
rs150400480
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:100102771 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGTTTCTCCCTTCAGCCTCCTAGAA[C/G]TGCTCTCCAGGTGAGGAGGGTTGGG
Phenotype
MIM: 602296 MIM: 600592 MIM: 612983 MIM: 612150 MIM: 612984 MIM: 602955
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
AP4M1 PubMed Links

Gene Details

Gene
AP4M1
Gene Name
adaptor related protein complex 4 mu 1 subunit
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004722.3 430 Silent Mutation CTG,GTG L82V NP_004713.2
XM_005250689.4 430 Missense Mutation CTG,GTG L89V XP_005250746.1
XM_005250690.4 430 Intron XP_005250747.1
XM_006716175.3 430 Missense Mutation CTG,GTG L89V XP_006716238.1
XM_011516685.2 430 Missense Mutation CTG,GTG L89V XP_011514987.1
XM_017012790.1 430 UTR 5 XP_016868279.1
XM_017012791.1 430 Intron XP_016868280.1
Gene
MCM7
Gene Name
minichromosome maintenance complex component 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001278595.1 430 Intron NP_001265524.1
NM_005916.4 430 Intron NP_005907.3
NM_182776.2 430 Intron NP_877577.1
XM_005250348.3 430 Intron XP_005250405.1
XM_017012217.1 430 Intron XP_016867706.1
Gene
MIR106B
Gene Name
microRNA 106b
There are no transcripts associated with this gene.

Gene
MIR25
Gene Name
microRNA 25
There are no transcripts associated with this gene.

Gene
MIR93
Gene Name
microRNA 93
There are no transcripts associated with this gene.

Gene
TAF6
Gene Name
TATA-box binding protein associated factor 6
There are no transcripts associated with this gene.

View Full Product Details