Product Details

SNP ID
rs150553593
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:103298635 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCTACCCAAGTTGTTCTGAATGTTC[C/G]TGAAACAAGAGTAACATGTTTAGAA
Phenotype
MIM: 603131
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
PMPCB PubMed Links

Gene Details

Gene
PMPCB
Gene Name
peptidase, mitochondrial processing beta subunit
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004279.2 201 Missense Mutation CCT,CGT P56R NP_004270.2
XM_005250717.3 201 Missense Mutation CCT,CGT P56R XP_005250774.1
XM_006716181.3 201 Missense Mutation CCT,CGT P56R XP_006716244.1

View Full Product Details