Product Details

SNP ID
rs115814584
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:144529467 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCCAGCAGCTCTGGCCCCGAGAAC[A/G]GACAGTCCCGGGCCCGGCGCCTTCC
Phenotype
MIM: 615880
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ARHGAP39 PubMed Links

Gene Details

Gene
ARHGAP39
Gene Name
Rho GTPase activating protein 39
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001308207.1 4566 UTR 3 NP_001295136.1
NM_001308208.1 4566 UTR 3 NP_001295137.1
NM_025251.2 4566 UTR 3 NP_079527.1
XM_011517308.1 4566 UTR 3 XP_011515610.1
XM_011517309.1 4566 UTR 3 XP_011515611.1
XM_011517312.2 4566 UTR 3 XP_011515614.1
XM_017013870.1 4566 UTR 3 XP_016869359.1
XM_017013871.1 4566 UTR 3 XP_016869360.1
Gene
C8orf82
Gene Name
chromosome 8 open reading frame 82
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001001795.1 4566 Intron NP_001001795.1
Gene
LRRC14
Gene Name
leucine rich repeat containing 14
There are no transcripts associated with this gene.

Gene
LRRC24
Gene Name
leucine rich repeat containing 24
There are no transcripts associated with this gene.

View Full Product Details