Product Details
- SNP ID
-
rs115821379
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.8:71198811 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TTTAAAAAAGTCTGGTGGGTGAATT[A/G]TACCTCAATAAAGCTGTTTTTTTAT
- Phenotype
-
MIM: 601653
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
EYA1
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs141811307] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- EYA1
- Gene Name
- EYA transcriptional coactivator and phosphatase 1
View Full Product Details