Product Details

SNP ID
rs117398242
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:67178075 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGCAAGGTACTTAGAGACAGGCCTG[A/G]CATATAAGTTTTATGTAAGTGTTTA
Phenotype
MIM: 604141 MIM: 611654
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ARFGEF1 PubMed Links

Gene Details

Gene
ARFGEF1
Gene Name
ADP ribosylation factor guanine nucleotide exchange factor 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006421.4 Intron NP_006412.2
XM_005251134.4 Intron XP_005251191.1
XM_005251135.3 Intron XP_005251192.1
XM_005251136.3 Intron XP_005251193.1
XM_011517442.2 Intron XP_011515744.1
Gene
CSPP1
Gene Name
centrosome and spindle pole associated protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001291339.1 Intron NP_001278268.1
NM_024790.6 Intron NP_079066.5
XM_005251305.3 Intron XP_005251362.2
XM_006716474.2 Intron XP_006716537.2
XM_006716477.2 Intron XP_006716540.2
XM_011517598.1 Intron XP_011515900.1
XM_011517599.1 Intron XP_011515901.1
XM_011517600.1 Intron XP_011515902.1
XM_011517601.1 Intron XP_011515903.1
XM_011517602.1 Intron XP_011515904.1
XM_011517603.1 Intron XP_011515905.1
XM_011517607.1 Intron XP_011515909.1
XM_011517608.1 Intron XP_011515910.1
XM_011517609.1 Intron XP_011515911.1
XM_011517611.2 Intron XP_011515913.1
XM_017013847.1 Intron XP_016869336.1
XM_017013848.1 Intron XP_016869337.1
XM_017013849.1 Intron XP_016869338.1
XM_017013850.1 Intron XP_016869339.1
XM_017013851.1 Intron XP_016869340.1
XM_017013852.1 Intron XP_016869341.1
XM_017013853.1 Intron XP_016869342.1
XM_017013854.1 Intron XP_016869343.1
XM_017013855.1 Intron XP_016869344.1
XM_017013856.1 Intron XP_016869345.1
XM_017013857.1 Intron XP_016869346.1
XM_017013858.1 Intron XP_016869347.1
XM_017013859.1 Intron XP_016869348.1

View Full Product Details