Product Details

SNP ID
rs139911017
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:142751146 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTCCGACACTCCTGTGGGCTCCGGA[C/T]AGACGTAGCAGCGCAGGGCCGGCGC
Phenotype
MIM: 606119
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LYPD2 PubMed Links

Gene Details

Gene
LYPD2
Gene Name
LY6/PLAUR domain containing 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_205545.2 166 Missense Mutation TAT,TGT Y28C NP_991108.1
Gene
SLURP1
Gene Name
secreted LY6/PLAUR domain containing 1
There are no transcripts associated with this gene.

View Full Product Details