Product Details

SNP ID
rs142508322
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:144309833 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGGACAGCGTCACCAAGCTGCTGA[C/T]GGACGTGCAGCTGATGAAGGGGAAG
Phenotype
MIM: 604900 MIM: 140580
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DGAT1 PubMed Links

Gene Details

Gene
DGAT1
Gene Name
diacylglycerol O-acyltransferase 1
There are no transcripts associated with this gene.

Gene
HSF1
Gene Name
heat shock transcription factor 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
XM_005272315.2 595 Missense Mutation ACG,ATG T142M XP_005272372.1
XM_005272316.2 595 Missense Mutation ACG,ATG T142M XP_005272373.1
XM_005272317.1 595 Missense Mutation ACG,ATG T142M XP_005272374.1
XM_011517004.2 595 UTR 5 XP_011515306.1
XM_011517006.2 595 Intron XP_011515308.1
XM_017013377.1 595 Missense Mutation ACG,ATG T142M XP_016868866.1
Gene
MIR6848
Gene Name
microRNA 6848
There are no transcripts associated with this gene.

View Full Product Details