Product Details
- SNP ID
-
rs143762729
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.8:10606895 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TGATTGTTTTCTAGCTTGATCTTGT[C/G]TAGAAATCTAAGTCATCTTGGCCAA
- Phenotype
-
MIM: 608581
- Polymorphism
- C/G, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
RP1L1
PubMed Links
Gene Details
- Gene
- RP1L1
- Gene Name
- retinitis pigmentosa 1-like 1
There are no transcripts associated with this gene.
View Full Product Details