Product Details

SNP ID
rs147098542
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:33548841 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTGTTACACATGGGGCAGACACAG[A/C]GAACTTCCAGCCATTTCACCAGACA
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
RNF122 PubMed Links

Gene Details

Gene
RNF122
Gene Name
ring finger protein 122
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_024787.3 785 Missense Mutation CGC,CTC R127L NP_079063.2
XM_017013846.1 785 Missense Mutation CGC,CTC R103L XP_016869335.1

View Full Product Details