Product Details

SNP ID
rs147334256
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:10607106 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTCCTCTCTGCATGAGGGGTCCCC[A/G]TGGACTTGGCATCAGGGCTCCTTGT
Phenotype
MIM: 608581
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
RP1L1 PubMed Links

Gene Details

Gene
RP1L1
Gene Name
retinitis pigmentosa 1-like 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_178857.5 7221 Missense Mutation ACG,ATG T2331M NP_849188.4

View Full Product Details