Product Details

SNP ID
rs147670749
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:20145327 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCTGGGAAAACCCGGGAAAGAACA[C/T]GTTGAGGAAAAGAGGGTAACACTCT
Phenotype
MIM: 193002
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SLC18A1 PubMed Links

Gene Details

Gene
SLC18A1
Gene Name
solute carrier family 18 member A1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001135691.2 2080 UTR 3 NP_001129163.1
NM_001142324.1 2080 UTR 3 NP_001135796.1
NM_001142325.1 2080 UTR 3 NP_001135797.1
NM_003053.3 2080 UTR 3 NP_003044.1
XM_011544625.1 2080 UTR 3 XP_011542927.1
XM_011544626.1 2080 UTR 3 XP_011542928.1
XM_011544627.1 2080 UTR 3 XP_011542929.1

View Full Product Details