Product Details

SNP ID
rs148302412
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:33549442 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAAAGGCGTGTTGGCACGGGAGCAC[A/G]CCTAACTCATCCTTCCCCTTGAAGT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
RNF122 PubMed Links

Gene Details

Gene
RNF122
Gene Name
ring finger protein 122
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_024787.3 726 Silent Mutation GGC,GGT G107G NP_079063.2
XM_017013846.1 726 Silent Mutation GGC,GGT G83G XP_016869335.1

View Full Product Details