Product Details

SNP ID
rs149753598
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:143614212 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCGGATTCCCTGTACCCCACACCGT[C/T]AGGGCCGAGCCGCTGCCTGCAGATT
Phenotype
MIM: 616408 MIM: 137020
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PYCRL PubMed Links

Gene Details

Gene
PYCRL
Gene Name
pyrroline-5-carboxylate reductase-like
There are no transcripts associated with this gene.

Gene
TSTA3
Gene Name
tissue specific transplantation antigen P35B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001317783.1 742 Silent Mutation CTA,CTG L211L NP_001304712.1
NM_003313.3 742 Silent Mutation CTA,CTG L205L NP_003304.1
XM_005251051.3 742 Silent Mutation CTA,CTG L205L XP_005251108.2
XM_011517269.1 742 Silent Mutation CTA,CTG L205L XP_011515571.1

View Full Product Details